EMAIL THIS PAGE TO A FRIEND
Arthritis research & therapy
Pathogenic roles of CXCL10 signaling through CXCR3 and TLR4 in macrophages and T cells: relevance for arthritis.
PMID 28724396
Abstract
Rheumatoid arthritis (RA) is a chronic autoimmune disease characterized by uncontrolled joint inflammation and destruction of bone and cartilage. We previously reported that C-X-C motif chemokine 10 (CXCL10; also called IP-10) has important roles in joint inflammation and bone destruction in arthritis. However, the specific mechanisms by which CXCL10 regulates the recruitment of inflammatory cells and the production of osteoclastogenic cytokines in RA progression are not fully understood. Bone marrow-derived macrophages and CD4 CXCL10 increased the migration of inflammatory cells through C-X-C chemokine receptor 3 (CXCR3)-mediated, but not toll-like receptor 4 (TLR4)-mediated, ERK activation. Interestingly, both receptors CXCR3 and TLR4 were simultaneously required for CXCL10-stimulated production of osteoclastogenic cytokines in CD4 These findings highlight the importance of CXCL10 signaling in the pathogenesis of RA and provide previously unidentified details of the mechanisms by which CXCL10 promotes the development of arthritis.