EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
TRIM37, a novel E3 ligase for PEX5-mediated peroxisomal matrix protein import.
PMID 28724525
Abstract
Most proteins destined for the peroxisomal matrix depend on the peroxisomal targeting signals (PTSs), which require the PTS receptor PEX5, whose deficiency causes fatal human peroxisomal biogenesis disorders (PBDs).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PEROX1 Peroxisome Isolation Kit, isolate peroxisomes from tissues and cells