EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Heterodimers of the transcriptional factors NFATc3 and FosB mediate tissue factor expression for 15(
PMID 28724635
Abstract
Tissue factor (TF) is expressed in vascular and nonvascular tissues and functions in several pathways, including embryonic development, inflammation, and cell migration. Many risk factors for atherosclerosis, including hypertension, diabetes, obesity, and smoking, increase TF expression. To better understand the TF-related mechanisms in atherosclerosis, here we investigated the role of 12/15-lipoxygenase (12/15-LOX) in TF expression. 15(