EMAIL THIS PAGE TO A FRIEND
Food & function
Fecal excretion of Maillard reaction products and the gut microbiota composition of rats fed with bread crust or bread crumb.
PMID 28725891
Abstract
A comparison between the impacts of advanced (N