EMAIL THIS PAGE TO A FRIEND
Aquatic toxicology (Amsterdam, Netherlands)
Accumulation and detoxification dynamics of Chromium and antioxidant responses in juvenile rare minnow, Gobiocypris rarus.
PMID 28728048
Abstract
Hexavalent chromium (Cr