EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Design, synthesis and evaluation of coumarin-pargyline hybrids as novel dual inhibitors of monoamine oxidases and amyloid-β aggregation for the treatment of Alzheimer's disease.
PMID 28728104
Abstract
A series of coumarin-pargyline hybrids (4a-x) have been designed, synthesized and evaluated as novel dual inhibitors of Alzheimer's disease (AD). Most of the compounds exhibited a potent ability to inhibit amyloid-β (Aβ) aggregation and monoamine oxidases. In particular, compound 4x exhibited remarkable inhibitory activities against monoamine oxidases (IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
126896
3-Fluorobenzylamine, 97%
C7H8FN