EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
Zebrafish have an ethanol-inducible hepatic 4-nitrophenol hydroxylase that is not CYP2E1-like.
PMID 28728133
Abstract
Zebrafish are an attractive model organism for toxicology; however, an important consideration in translating between species is xenobiotic metabolism/bioactivation. CYP2E1 metabolizes small hydrophobic molecules, e.g. ethanol, cigarette smoke, and diesel exhaust components. CYP2E1 is thought to only be conserved in mammals, but recent reports identified homologous zebrafish cytochrome P450s. Herein, ex vivo biochemical measurements show that unlike mammals, zebrafish possess a low-affinity 4-nitrophenol hydroxylase (K
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
222569
Fomepizole, 99%
C4H6N2