EMAIL THIS PAGE TO A FRIEND
The EMBO journal
Cohesins and condensins orchestrate the 4D dynamics of yeast chromosomes during the cell cycle.
PMID 28729434
Abstract
Duplication and segregation of chromosomes involves dynamic reorganization of their internal structure by conserved architectural proteins, including the structural maintenance of chromosomes (SMC) complexes cohesin and condensin. Despite active investigation of the roles of these factors, a genome-wide view of dynamic chromosome architecture at both small and large scale during cell division is still missing. Here, we report the first comprehensive 4D analysis of the higher-order organization of the