EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
A Novel and Validated Protocol for Performing MIC Tests to Determine the Susceptibility of
PMID 28729865
Abstract
This paper presents a validated protocol, using a novel, specifically formulated medium, to perform broth microdilution antimicrobial susceptibility assays of the salmonid bacterial pathogen
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F1427
Florfenicol, analytical standard, for drug analysis
C12H14Cl2FNO4S