EMAIL THIS PAGE TO A FRIEND
Clinical and experimental pharmacology & physiology
Effects of nitric oxide on large-conductance Ca
PMID 28731589
Abstract
The human cardiac fibroblast (HCF) is the most abundant cell type in the myocardium, and HCFs play critical roles in maintaining normal cardiac function. However, unlike cardiomyocytes, the electrophysiology of HCFs is not well established. In the cardiovascular system, Ca