EMAIL THIS PAGE TO A FRIEND
Bone
Rad GTPase is essential for the regulation of bone density and bone marrow adipose tissue in mice.
PMID 28732776
Abstract
The small GTP-binding protein Rad (RRAD, Ras associated with diabetes) is the founding member of the RGK (Rad, Rem, Rem2, and Gem/Kir) family that regulates cardiac voltage-gated Ca