EMAIL THIS PAGE TO A FRIEND
Ultrasonics sonochemistry
Sonochemical synthesis, formation mechanism, and solar cell application of tellurium nanoparticles.
PMID 28732940
Abstract
Tellurium inherently tends to form 1-D structures and while the 0-D Te nanostructures have better properties and applications in solar cell. In the present study, 0-D Te nanostructures including rice-like and spherical nanoparticles with the particle size of 15-40nm were successfully synthesized via a facile sonochemical method. In the absence of ultrasonic irradiation nanorods were produced while performing the reaction under ultrasonic waves (at 200W for 30min) led to the formation of nanoparticles. Finally, the efficiency of various as-synthesized Te nanostructures in quantum dot-sensitized solar cells (QDSSCs) were evaluated. Using rice-like nanoparticles led to increase in J
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
155098
Valeronitrile, 99.5%
C5H9N