EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
ANGPTL4 promotes bile acid absorption during taurocholic acid supplementation via a mechanism dependent on the gut microbiota.
PMID 28733267
Abstract
Angiopoietin-like 4 (ANGPTL4) raises plasma triglyceride levels by inhibiting lipoprotein lipase. A set of compounds that are able to reduce plasma triglyceride levels are bile acids (BA). Because BA have been shown to decrease ANGPTL4 secretion by intestinal cells, we hypothesized that BA lower plasma triglycerides (partly) via ANGPTL4. To test that hypothesis, wild-type and Angptl4