EMAIL THIS PAGE TO A FRIEND
Reproduction (Cambridge, England)
Regulation and action of early growth response 1 in bovine granulosa cells.
PMID 28733346
Abstract
Fibroblast growth factors (FGF) modify cell proliferation and differentiation through receptor tyrosine kinases, which stimulate the expression of transcription factors including members of the early growth response (EGR) family. In ovarian granulosa cells, most FGFs activate typical response genes, although the role of EGR proteins has not been described. In the present study, we determined the regulation of EGR mRNA by FGFs and explored the role of EGR1 in the regulation of FGF-response genes. Addition of FGF1, FGF2, FGF4 or FGF8b increased