EMAIL THIS PAGE TO A FRIEND
Proteins
A computational method for selecting short peptide sequences for inorganic material binding.
PMID 28734030
Abstract
Discovering or designing biofunctionalized materials with improved quality highly depends on the ability to manipulate and control the peptide-inorganic interaction. Various peptides can be used as assemblers, synthesizers, and linkers in the material syntheses. In another context, specific and selective material-binding peptides can be used as recognition blocks in mining applications. In this study, we propose a new in silico method to select short 4-mer peptides with high affinity and selectivity for a given target material. This method is illustrated with the calcite (104) surface as an example, which has been experimentally validated. A calcite binding peptide can play an important role in our understanding of biomineralization. A practical aspect of calcite is a need for it to be selectively depressed in mining sites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS000645
Dolomite, AldrichCPR
C2CaMgO6