EMAIL THIS PAGE TO A FRIEND
The Journal of chemical physics
Effect of interlamellar interactions on shear induced multilamellar vesicle formation.
PMID 28734290
Abstract
Shear-induced multilamellar vesicle (MLV) formation has been studied by coupling the small-angle neutron scattering (SANS) technique with neutron spin echo (NSE) spectroscopy. A 10% mass fraction of the nonionic surfactant pentaethylene glycol dodecyl ether (C
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76437
Pentaethylene glycol monododecyl ether, BioXtra, ≥98.0% (GC)
C22H46O6