EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Dried urine spots - A novel sampling technique for comprehensive LC-MS
PMID 28734350
Abstract
Dried matrix spot (DMS) technique as alternative sampling strategy, especially dried urine spots (DUS), might be an alternative for drug screening. So far only particular drugs or drug classes were covered in DMS screenings. Therefore, workup of DUS for a broad comprehensive library-based LC-MS
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50787
Bisoprolol, analytical standard
C18H31NO4