EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Streptavidin-biotin-peroxidase nanocomplex-amplified microfluidics immunoassays for simultaneous detection of inflammatory biomarkers.
PMID 28734353
Abstract
Simultaneous, sensitive and quantitative detection of biomarkers in infectious disease is crucial for guiding antimicrobial treatment and predicting prognosis. This work reported an ultrasensitive and quantitative microfluidic immunoassay combined with the streptavidin-biotin-peroxidase (SA-B-HRP) nanocomplex-signal amplification system (MIS) to detect two inflammatory biomarkers, procalcitonin (PCT, for discriminating bacterial infections from nonbacterial infections) and interleukin-6 (IL-6, for monitoring the kinetics of infectious disease) simultaneously. The amplification system was based on the one step self-assembly of SA and B-HRP to form the SA-B-HRP nanocomplex, which effectively amplified the chemiluminescent signals. The linear ranges for PCT and IL-6 detections by MIS were 250-1.28 × 10