EMAIL THIS PAGE TO A FRIEND
Talanta
Development of a solid-phase extraction method with simple MEKC-UV analysis for simultaneous detection of indole metabolites in human urine after administration of indole dietary supplement.
PMID 28738586
Abstract
This work presents the development of a solid phase extraction method with simple MEKC-UV analysis for the simultaneous determination of indole-3-carbinol (I3C) and its metabolites (3, 3'-diindolylmethane (DIM), indole-3-carboxaldehyde (I3CAL), indole-3-acetonitrile (I3A)) in human urine after oral administration of an indole dietary supplement. Solid phase extraction (SPE) method was applied for the first time for simultaneous analysis of these indole metabolites. The MEKC separation method was developed in a previous work. Three commercial SPE cartridges, each with different sorbent materials, were investigated: Sep-Pak