EMAIL THIS PAGE TO A FRIEND
Brain research
Inhibition by neosurugatoxin of nicotine-induced antinociception.
PMID 2873866
Abstract
We have found a selective inhibition of nicotine-induced antinociception in mice by neosurugatoxin (NSTX, 0.4-3.8 nmol/kg), a neurotoxin with a high affinity for ganglionic nicotinic receptors (ED50 = 0.65 nmol/kg). The toxin also reduced specific [3H]nicotine binding in mouse brain membranes (IC50 = 95 nM). The anti-nicotinic activity of NSTX was markedly greater than that of mecamylamine and pempidine. These data indicate that NSTX may block functional nicotinic cholinoceptors possibly in the central nervous system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274631
1,2,2,6,6-Pentamethylpiperidine, 97%
C10H21N