EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
CXCL1 and CXCL2 Regulate NLRP3 Inflammasome Activation via G-Protein-Coupled Receptor CXCR2.
PMID 28739876
Abstract
Inflammation is an extensively concerted process that confers protection to the host encountering immune insult. The major inflammatory mediators include IL-1 family members, such as IL-1β, and the functional activation of such molecules is arbitrated by their regulated cleavage brought about by components of a multiprotein complex called inflammasome. In this context, NLR family pyrin domain containing 3 (NLRP3) inflammasome activation often acts as a rate-limiting step in regulating critical cell-fate decisions in various inflammatory scenarios. In this study, we identify the G-protein-coupled receptor CXCR2 (recognizing chemokines CXCL1 and CXCL2) as another arm feeding into the regulated activation of NLRP3 inflammasome in macrophages. We demonstrate that in vivo blocking of CXCL1 and CXCL2 can significantly reduce the
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0280
CP-456773 sodium salt, ≥98% (HPLC)
C20H23N2NaO5S