EMAIL THIS PAGE TO A FRIEND
Nature communications
Spatial cycles mediated by UNC119 solubilisation maintain Src family kinases plasma membrane localisation.
PMID 28740133
Abstract
The peripheral membrane proto-oncogene Src family protein tyrosine kinases relay growth factor signals to the cytoplasm of mammalian cells. We unravel the spatial cycles of solubilisation, trapping on perinuclear membrane compartments and vesicular transport that counter entropic equilibration to endomembranes for maintaining the enrichment and activity of Src family protein tyrosine kinases at the plasma membrane. The solubilising factor UNC119 sequesters myristoylated Src family protein tyrosine kinases from the cytoplasm, enhancing their diffusion to effectively release Src family protein tyrosine kinases on the recycling endosome by localised Arl2/3 activity. Src is then trapped on the recycling endosome via electrostatic interactions, whereas Fyn is quickly released to be kinetically trapped on the Golgi by palmitoyl acyl-transferase activity. Vesicular trafficking from these compartments restores enrichment of the Src family protein tyrosine kinases to the plasma membrane. Interference with these spatial cycles by UNC119 knockdown disrupts Src family protein tyrosine kinase localisation and signalling activity, indicating that UNC119 could be a drug target to affect oncogenic Src family protein tyrosine kinase signalling.The peripheral membrane proto-oncogene Src family protein tyrosine kinases (SFKs) transmit growth factor signals to the cytoplasm. Here the authors show that the solubilising factor UNC119 sequesters myristoylated SFKs to maintain its enrichment at the plasma membrane to enable signal transduction.