EMAIL THIS PAGE TO A FRIEND
Journal of food science and technology
Variation of active constituents and antioxidant activity in
PMID 28740285
Abstract
It has been claimed that inflorescences of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00740580
Quercitrin, primary reference standard
C21H20O11