EMAIL THIS PAGE TO A FRIEND
Journal of food science and technology
Improving oxidative stability of virgin olive oil by addition of microalga
PMID 28740304
Abstract
Antioxidant activity of Chlorella (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A88255
p-Anisidine, ≥99%
C7H9NO