EMAIL THIS PAGE TO A FRIEND
Scientific reports
Lithium Sensitive ORAI1 Expression, Store Operated Ca
PMID 28743945
Abstract
Chorea-Acanthocytosis (ChAc), a neurodegenerative disorder, results from loss-of-function-mutations of chorein-encoding gene VPS13A. In tumour cells chorein up-regulates ORAI1, a Ca