EMAIL THIS PAGE TO A FRIEND
Teratogenesis, carcinogenesis, and mutagenesis
Prenatal exposure to the fungicide dinocap causes behavioral torticollis, ballooning and cleft palate in mice, but not rats or hamsters.
PMID 2874635
Abstract
The present study is an evaluation of the developmental toxicity of dinocap in three rodent species using an in vivo teratology screen. Our protocol uses postnatal viability, weight gain, and morphological and behavioral development through weaning to assess the developmental toxicity of compounds. Dinocap administered orally on days 7 to 16 of gestation to the CD-1 mouse resulted in increased postnatal mortality at 25 mg/kg/d (80% in block 1 and 40% in block 2). Many of the treated pups that died during the neonatal period were "ballooned" and had cleft palates. Although there was no treatment related mortality in the 12 mg/kg/d dosage group, 6% (14/226) of these mice and 24% (23/96) of the survivors from the 25 mg/kg/d dosage group displayed torticollis (a twisting of the neck resulting in an abnormal tilting of the head). These tilted-head mice held the head and forepart of the body tilted constantly to one side, both when resting and walking. The tilt was in either direction but was always constant for a given animal; in different mice, the angle varied considerably from almost 0 to 30 degrees. Some mice circled repeatedly in one direction in the home cage, others bobbed their heads and did back-flips, while others rolled over, always rolling in the same direction. In the hamster, developmental toxicity was seen at (100 and 200 mg/kg/d) or near (50 mg/kg/d) maternally toxic doses but no behavioral alterations were noted and none of the pups were ballooned.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45452
Dinocap, PESTANAL®, analytical standard
C18H24N2O6