EMAIL THIS PAGE TO A FRIEND
Arquivos brasileiros de cardiologia
Single Derivation Fragmented QRS Can Predict Poor Prognosis in Successfully Revascularized Acute STEMI Patients.
PMID 28746519
Abstract
QRS fragmentation (fQRS) is classically defined as the presence of slurred QRS morphology in at least two contiguous leads, and its prognostic importance has been shown in ST elevation myocardial infarction (STEMI). However, no study has investigated the significance of single lead fQRS (sl-fQRS) in surface electrocardiography (ECG). To evaluate whether sl-fQRS is as valuable as classical fQRS in patients with acute STEMI who had successful revascularization with primary percutaneous coronary intervention (pPCI). We included 330 patients with a first STEMI who had been successfully revascularized with pPCI. The patient's electrocardiography was obtained in the first 48 hours, and the patients were divided into three groups according to the absence of fQRS (no-fQRS); fQRS presence in a single lead (sl-fQRS); and ≥2 leads with fQRS (classical fQRS). In-hospital mortality was significantly higher both in patients with sl-fQRS and in patients with ≥ 2 leads with fQRS compared to patients with no-fQRS. In ROC curve analysis, ≥ 1 leads with fQRS yielded a sensitivity of 75% and specificity of 57.4% for the prediction of in-hospital mortality. Multivariate analysis showed that sl-fQRS is an independent predictor of in-hospital mortality (OR: 3.989, 95% CI: 1.237-12.869, p = 0.021). Although the concept of at least two derivations is mentioned for the classical definition of fQRS, our study showed that fQRS in only one lead is also associated with poor outcomes. Therefore, ≥1 leads with fQRS can be useful when describing the patients under high cardiac risk in acute STEMI.