EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Comparison between several reactors with Trametes versicolor immobilized on lignocellulosic support for the continuous treatments of hospital wastewater.
PMID 28746994
Abstract
Hospital wastewater is a major source of pharmaceutically active compounds (PhACs), which are not all removed in conventional wastewater treatment plants. White rot fungi can degrade PhACs, but their application has been limited to non-sterile conditions due to the competition with other microorganisms for growth. In this study, immobilization of Trametes versicolor on different lignocellulosic supports was studied as strategy to ensure fungal survival under continuous treatment conditions. A fluidized bed reactor and a trickling packed-bed reactor with T. versicolor immobilized on pallet wood were employed for the removal of ibuprofen, ketoprofen and naproxen. Best results were obtained with the trickling packed-bed reactor, which operated for 49days with high removal values in real hospital wastewater.