EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Identification of a novel component leading to anti-tumor activity besides the major ingredient cordycepin in Cordyceps militaris extract.
PMID 28750234
Abstract
In accordance with our previous study that was carried out to identify novel anti-tumor ingredients, chromatographic separation in combination with an anti-tumor activity assay was used for analysis of Cordyceps militaris extract in this study. Various modes of chromatography including reversed-phase, cation-exchange and anion-exchange were used to separate components of Cordyceps militaris, which showed various chemical properties. Anti-tumor activity of each fraction was assessed by a Hoechst staining-based apoptosis assay using malignant melanoma MeWo cells. By these repeated approaches through chromatographic segregation and cell biological assay, we finally succeeded in identifying the target substance from a certain fraction that included neutral hydrophilic components using a pre-column and post-column chlorine adduct ionization LC-APCI-MS method. The target substance was a mono-carbohydrate, xylitol, that induced apoptotic cell death in MeWo cells but not in normal human OUMS-24 fibroblasts. This is the first study showing that Cordyceps militaris extract contains a large amount of xylitol. Thus, our results will contribute greatly to uncovering the mysterious multifunctional herbal drug Cordyceps militaris as an anti-tumor agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
86265
D-(+)-Talose, ≥99.0% (HPLC)
C6H12O6
56224
L-(+)-Gulose, ≥98.0% (HPLC)
C6H12O6