EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
MoS
PMID 28750338
Abstract
Stimuli-responsive carriers have extensively attracted attention in recent years. However, long-term and real-time tracking ability with stimuli-responsive carrier is still in its infant stage due to the limitations such as, low efficacy, instability and cytotoxicity in a bio-environment. In this work, we developed a reduction-sensitive carrier composed of lipoic acid-modified low molecular weight polyethyleneimine (LA-PEI) and large surface ratio MoS2 nanosheet integrated via disulfide bond to mimic a high molecular weight PEI. The positively charged carriers loading negatively charged aptamer enter the cells for a real time long-term tracking of adenosine triphosphate (ATP) metabolism in glioma stem cells (GSCs) when stimulated by TGFβ factor secreted from HUVECs. We envision that MoS