EMAIL THIS PAGE TO A FRIEND
Experimental and clinical endocrinology & diabetes : official journal, German Society of Endocrinology [and] German Diabetes Association
Hyperthyroidism in Patients with Graves' Ophthalmopathy, and Thyroidal, Skeletal and Eye Muscle Specific Type 2 Deiodinase Enzyme Activities.
PMID 28750432
Abstract
Graves' ophthalmopathy is characterized by hyperthyroidism, which is associated with higher serum T