EMAIL THIS PAGE TO A FRIEND
PMID 28750485
Abstract
The increased resistance of We screened a small molecule library-the library of pharmacologically active compounds (LOPAC), which includes 1,280 pharmacologically active compounds-to identify inhibitors of We identified diphenyleneiodonium (DPI) as a novel anti- DPI may be a candidate anti-