EMAIL THIS PAGE TO A FRIEND
Cellular immunology
Didox (3,4-dihydroxybenzohydroxamic acid) suppresses IL-33-induced cytokine production in primary mouse mast cells.
PMID 28750923
Abstract
While IgE is considered the primary mediator of mast cell activation, IL-33 contributes substantially in asthma, allergic rhinitis, and atopic dermatitis. To develop effective treatments for allergic disease, it is important to understand the role of therapeutic agents on IL-33 activation. We examined the effect of Didox (3,4-dihydroxybenzohydroxamic acid), an antioxidant and ribonucleotide reductase (RNR) inhibitor, on IL-33-mediated mast cell activation. Didox suppressed IL-6, IL-13, TNF, and MIP-1α (CCL3) production in bone marrow derived mast cells following IL-33 activation. This suppression was observed in different genetic backgrounds and extended to peritoneal mast cells. The antioxidant N-acetylcysteine mimicked the suppression of Didox, albeit at a much higher dose, while the RNR inhibitor hydroxyurea had no effect. Didox substantially suppressed IL-33-mediated NFκB and AP-1 transcriptional activities. These results suggest that Didox attenuates IL-33-induced mast cell activation and should be further studied as a potential therapeutic agent for inflammatory diseases involving IL-33.