EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
A lipodystrophy-causing lamin A mutant alters conformation and epigenetic regulation of the anti-adipogenic
PMID 28751304
Abstract
Mutations in the