Biology open
EsGLUT4 and CHHBP are involved in the regulation of glucose homeostasis in the crustacean Eriocheir sinensis.
PMID 28751307
Glucose is an essential energy source for both vertebrates and invertebrates. In mammals, glucose uptake is mediated primarily by glucose transporters (GLUTs), members of the major facilitator superfamily (MFS) of passive transporters. Among the GLUTs, GLUT4 is the main glucose transporter in muscles and adipocytes. In skeletal muscle cells, GLUT4 interacts with the lipid raft protein flotillin to transport glucose upon stimulation by insulin. Although several studies have examined GLUT4 function in mammals, few have been performed in crustaceans, which also use glucose as their main energy source. Crustacean hyperglycemic hormone (CHH) is a multifunctional neurohormone found only in arthropods, and one of its roles is to regulate glucose homeostasis. However, the molecular mechanism that underlies CHH regulation and whether GLUT4 is involved in its regulation in crustaceans remain unclear. In the present study, we identified a full-length GLUT4 cDNA sequence (defined herein as EsGLUT4) from the Chinese mitten crab Eriocheir sinensis and analyzed its tissue distribution and cellular localization. By the ForteBio Octet system, two large hydrophilic regions within EsGLUT4 were found to interact with the CHH binding protein (CHHBP), an E. sinensis flotillin-like protein. Interestingly, live-cell imaging indicated that EsGLUT4 and CHHBP responded simultaneously upon stimulation by CHH, resulting in glucose release. In contrast to insulin-dependent GLUT4, however, EsGLUT4 and CHHBP were present within cytoplasmic vesicles, both translocating to the plasma membrane upon CHH stimulation. In conclusion, our results provide new evidence for the involvement of EsGLUT4 and CHHBP in the regulation of glucose homeostasis in crustacean carbohydrate metabolism.