EMAIL THIS PAGE TO A FRIEND
Molecular cancer research : MCR
Selective MET Kinase Inhibition in MET-Dependent Glioma Models Alters Gene Expression and Induces Tumor Plasticity.
PMID 28751462
Abstract
The receptor tyrosine kinase (RTK) MET represents a promising tumor target in a subset of glioblastomas. Most RTK inhibitors available in the clinic today, including those inhibiting MET, affect multiple targets simultaneously. Previously, it was demonstrated that treatment with cabozantinib (MET/VEGFR2/RET inhibitor) prolonged survival of mice carrying orthotopic patient-derived xenografts (PDX) of the MET-addicted glioblastoma model E98, yet did not prevent development of recurrent and cabozantinib-resistant tumors. To exclude VEGFR2 inhibition-inflicted blood-brain barrier normalization and diminished tumor distribution of the drug, we have now investigated the effects of the novel MET-selective inhibitor Compound A in the orthotopic E98 xenograft model.