EMAIL THIS PAGE TO A FRIEND
EMBO molecular medicine
Promoterless gene targeting without nucleases rescues lethality of a Crigler-Najjar syndrome mouse model.
PMID 28751579
Abstract
Crigler-Najjar syndrome type I (CNSI) is a rare monogenic disease characterized by severe neonatal unconjugated hyperbilirubinemia with a lifelong risk of neurological damage and death. Liver transplantation is the only curative option, which has several limitations and risks. We applied an