EMAIL THIS PAGE TO A FRIEND
ChemSusChem
Broadband-Emitting 2 D Hybrid Organic-Inorganic Perovskite Based on Cyclohexane-bis(methylamonium) Cation.
PMID 28752545
Abstract
A new broadband-emitting 2 D hybrid organic-inorganic perovskite (CyBMA)PbBr
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
180467
1,3-Cyclohexanebis(methylamine), mixture of isomers, 98%
C8H18N2