EMAIL THIS PAGE TO A FRIEND
Domestic animal endocrinology
Ovarian steroids, oxytocin, and tumor necrosis factor modulate equine oviduct function.
PMID 28753494
Abstract
The oviduct plays important roles in the early reproductive process. The aim of this study was to evaluate gene transcription and protein expression of progesterone receptor (PGR), estrogen receptors 1 (ESR1) and 2 (ESR2); oxytocin receptor (OXTR); prostaglandin F2α synthase (AKR1C3), and prostaglandin E2 synthase (Ptges) in mare oviduct in different estrous cycle stages. Estradiol (E