EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
CaMKII inhibition mitigates ischemia/reperfusion-elicited calpain activation and the damage to membrane skeleton proteins in isolated rat hearts.
PMID 28754591
Abstract
Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2142
KN-62, ≥95%, powder
C38H35N5O6S2
K1385
KN-93, ≥98% (HPLC)
C26H29ClN2O4S · H3PO4