EMAIL THIS PAGE TO A FRIEND
Acta biomaterialia
Biomimetically grown apatite spheres from aggregated bioglass nanoparticles with ultrahigh porosity and surface area imply potential drug delivery and cell engineering applications.
PMID 28754647
Abstract
Here we communicate the generation of biomimetically grown apatite spheres from aggregated bioglass nanoparticles and the potential properties applicable for drug delivery and cell/tissue engineering. Ion releasing nanoparticulates of bioglass (85%SiO This work reports the generation of apatite spheres with a few micrometers in size biomimetically grown from bioactive glass nanoparticles, through a series of intriguing yet unprecedented phenomenon involving aggregation of nanoparticles, mineralization and sphere growth. The mineral microspheres possess some unique physico-chemical properties including mesoporosity, ultrahigh surface area, and therapeutic ionic release. Furthermore, the spheres show excellent loading and delivery capacity of protein molecules, and mediate favorable cellular interactions in 2D and 3D culture conditions, demonstrating a future multifunctional microcarrier platform for the therapeutics delivery and cell/tissue engineering.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3912
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%