EMAIL THIS PAGE TO A FRIEND
PMID 28754720
Abstract
There is a high demand for noninvasive screening tools for gastrointestinal cancer (GIC) detection, and GIC-specific markers are required for such purposes. It is established that induction of the telomerase reverse transcriptase gene ( Sixty-nine GIC patients (34 colorectal carcinoma and 35 gastric cancer) and 62 healthy adults were recruited and fecal samples were collected. Paired tumors and adjacent non-cancerous tissues from 34 patients and normal mucosa tissues from 12 healthy individuals were collected. We identified two GIC-specific methylation sites at -218 (CpG site 1) and -210 (CpG site 2) in the This proof-of-concept study suggests the feasibility of stool Induction of telomerase reverse transcriptase (TERT) expression coupled with telomerase activation is essential for cancer development/progression, while aberrant TERT promoter methylation has been linked to TERT induction in oncogenesis. We identified two cancer-specific methylation sites (CpG1 and 2) in the TERT promoter in tumors from GIC patients. Methylated TERT promoter CpG sites 1 and 2 were detectable in patient stool, while only background levels were observed in healthy individuals. The sensitivity and specificity was comparable to other known stool methylation markers for GIC detection. This proof-of-concept study suggests the feasibility of stool TERT promoter methylation analyses for noninvasive screening of GIC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
364959
N-[Tris(hydroxymethyl)methyl]acrylamide, contains ≤7% KCl, 93%
C7H13NO4