EMAIL THIS PAGE TO A FRIEND
Journal of photochemistry and photobiology. B, Biology
Green synthesized nickel nanoparticles for targeted detection and killing of S. typhimurium.
PMID 28755580
Abstract
Simple and sensitive colorimetric immunosensor based on peroxidase mimetic activity and photothermal effect of nickel oxide nanoparticle (NiOGs) has been developed to detect and kill food borne pathogen Salmonella typhimurium. NiOGs showed superior peroxidase mimetic activity for oxidation of peroxidase substrate 3, 3', 5, 5'-tetramethylbenzidine (TMB). Oxidation of TMB by NiOGs followed Michaelis-Menten kinetics with K