EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
Molecular and biochemical characterisation and recognition by the immune host of the glyceraldehyde 3-phosphate dehydrogenase (GAPDH) of the abomasal nematode parasite Teladorsagia circumcincta.
PMID 28757123
Abstract
A 1023 bp full length cDNA encoding Teladorsagia circumcincta GAPDH (TeciGAPDH) was cloned, expressed in Escherichia coli and the recombinant protein purified and its kinetic properties determined. A phylogenetic tree was constructed using helminth GAPDH sequences. The predicted protein consisted of 341 amino acids and was present as a single band of about 38 kDa on SDS-PAGE. Multiple alignments of the protein sequence of TeciGAPDH with homologues from other helminths showed that the greatest similarity (93%) to the GAPDH of Haemonchus contortus and Dictyocaulus viviparus, 82-86% similarity to the other nematode sequences and 68-71% similarity to cestode and trematode enzymes. Substrate binding sites and conserved regions were identified and were completely conserved in other homologues. At 25 °C, the optimum pH for TeciGAPDH activity was pH 8, the V