EMAIL THIS PAGE TO A FRIEND
Nature neuroscience
Zika virus directly infects peripheral neurons and induces cell death.
PMID 28758997
Abstract
Zika virus (ZIKV) infection is associated with neurological disorders of both the CNS and peripheral nervous systems (PNS), yet few studies have directly examined PNS infection. Here we show that intraperitoneally or intraventricularly injected ZIKV in the mouse can infect and impact peripheral neurons in vivo. Moreover, ZIKV productively infects stem-cell-derived human neural crest cells and peripheral neurons in vitro, leading to increased cell death, transcriptional dysregulation and cell-type-specific molecular pathology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6546
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%
C10H12O4