EMAIL THIS PAGE TO A FRIEND
Water research
Iodinated trihalomethane formation during chloramination of iodate-containing waters in the presence of zero valent iron.
PMID 28759794
Abstract
Iodide (I
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
281409
2-Iodophenol, 98%
C6H5IO