EMAIL THIS PAGE TO A FRIEND
Chemosphere
High proportions of inorganic arsenic in Laminaria digitata but not in Ascophyllum nodosum samples from Ireland.
PMID 28759813
Abstract
Seaweed can accumulate inorganic arsenic (iAs) from seawater as hydrogen arsenate (HAsO