EMAIL THIS PAGE TO A FRIEND
The Journal of experimental biology
A mosquito entomoglyceroporin,
PMID 28760831
Abstract
The mosquito
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63052
Magnesium acetate solution, BioUltra, for molecular biology, ~1 M in H2O
C4H6MgO4