EMAIL THIS PAGE TO A FRIEND
Journal of cell science
The cholesterol ester cycle regulates signalling complexes and synapse damage caused by amyloid-β.
PMID 28760925
Abstract
Cholesterol is required for the formation and function of some signalling platforms. In synaptosomes, amyloid-β (Aβ) oligomers, the causative agent in Alzheimer's disease, bind to cellular prion proteins (PrP