EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Elevated auxin biosynthesis and transport underlie high vein density in C
PMID 28761000
Abstract
High vein density, a distinctive trait of C